Learn More
Cole-Parmer Adhering Mat, Blue, 18" x 18" for Orbital- Shaker
Catalog No. NC0707840 Shop All Cole-Parmer Products
Encompass_Preferred
Scientific Industries Genie SI-1617 Adhering Mat, Blue, 18" x 18" for Orbital- Shaker, Enhance the versatility of your Scientific Industries Orbital Shaker, Mat is naturally adhering and perfect for use with open containers. Rinse clean to renew adhering properties.
Catalog No. NC0707840 Supplier Cole-Parmer Supplier No. 5182108
Scientific Industries Genie SI-1617 Adhering Mat, Blue, 18" x 18" for Orbital- Shaker, Enhance the versatility of your Scientific Industries Orbital Shaker, Mat is naturally adhering and perfect for use with open containers. Rinse clean to renew adhering properties.