Learn More
MATSUNAMI GLASS USA ADHESION MICROSCOPE SLIDES
Catalog No. NC1889570
Encompass_Preferred
TOMO® Adhesion Microscope Slides. Matsunami TOMO® hydrophilic adhesion slides offer superior tissue adhesion, staining performance and slide surface fluidics. Proprietary slide coating ensures consistent performance even after exposure to heat stress and humid environmental conditions. Optimized for use with thermal printers.Recommended for use on Ventana® Benchmark staining platform and automated IHC platforms.Matsunami microscope slides are manufactured in Japan using optically clear, exceptionally flat, low fluorescence glass and finished with 90° and clipped corners.Slides are packaged 100 per box, 10 boxes per case (1000 slides).
This item is not returnable. View return policy
Catalog No. NC1889570 Supplier MATSUNAMI GLASS USA Supplier No. TOM11/90
TOMO® Adhesion Microscope Slides. Matsunami TOMO® hydrophilic adhesion slides offer superior tissue adhesion, staining performance and slide surface fluidics. Proprietary slide coating ensures consistent performance even after exposure to heat stress and humid environmental conditions. Optimized for use with thermal printers.Recommended for use on Ventana® Benchmark staining platform and automated IHC platforms.Matsunami microscope slides are manufactured in Japan using optically clear, exceptionally flat, low fluorescence glass and finished with 90° and clipped corners.Slides are packaged 100 per box, 10 boxes per case (1000 slides).