Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Selectfrost™ Adhesion Slides

Catalog No. 22037291 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Aqua
Blue
Green
Lilac
Pink
Tan
White
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-037-291 Green
22-037-294 Aqua
22-037-290 Blue
22-037-301 Lilac
22-037-292 Pink
22-037-302 Tan
22-037-200 White
22-037-293 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 22-037-291
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fisherbrand™ Selectfrost™ Adhesion Slides are available in a variety of colors.

Fisherbrand™ Selectfrost™ Adhesion Slides are available in a variety of colors.

  • Special treatment process that permits tissue samples/cells adhere to the slide without additional adhesive. 
  • Autoclavable 
  • Available in an array of tab colors 
  • Durable, opaque tab is inert to common chemicals and reagents 
  • Includes adhesion marking to distinguish orientation 
  • Packaged in paperboard box 
  • Size 25 x 75 x 1mm 

Specifications

Length (Metric) 75 mm
Frosted Selectfrost Color-coded
Type Microscope Slide
Width (Metric) 25 mm
Color Green
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.