Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences HybriWell™ Sealing System Chamber 3 well 100 mcL/Chbr, 20mm x 21mm
SDP

Catalog No. 5026622
Encompass_Preferred

HybriWell Sealing Chambers System seals to a microscope slide surface enclosing a single or multiple specimens in a small reagent volume. They are suitable for most protein and nucleic acid assays. Access ports in the chamber surface allow for the addition or removal of reactants and posrts are easily sealed using adhesive seal tabs (provided) to prevent reagent evaporation. These sealed chambers are watertight and can be used for water-bath incubations. RNase free, hydrophobic surfaces will not trap or bind probes. Disposable chamber remove cleanly and easily even after heating. Coms with non-adhesive peeling tab.
Volume: 100 µl/well
Chamber: 20mm x 21mm
Depth: 0.15mm
O.D.: 25mm x 75mm

Catalog No. 50-266-22 Supplier Electron Microscopy Sciences Supplier No. 7032830
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HybriWell Sealing Chambers System seals to a microscope slide surface enclosing a single or multiple specimens in a small reagent volume. They are suitable for most protein and nucleic acid assays. Access ports in the chamber surface allow for the addition or removal of reactants and posrts are easily sealed using adhesive seal tabs (provided) to prevent reagent evaporation. These sealed chambers are watertight and can be used for water-bath incubations. RNase free, hydrophobic surfaces will not trap or bind probes. Disposable chamber remove cleanly and easily even after heating. Coms with non-adhesive peeling tab.
Volume: 100 µl/well
Chamber: 20mm x 21mm
Depth: 0.15mm
O.D.: 25mm x 75mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.