Learn More
Research Products International Corp Plate Sealing Films, ThermalSeal, Clear, Non-Sterile, 100 per Package
SDP
Catalog No. 50550228
Encompass
Adhesive films are specially designed for use as microplate covers. Prevent sample evaporation and contamination in sensitive PCR applications. RNase/DNase free. It is thermostable and functional from -40°C to +120°C at 75% humidity. ThermalSeal is a Trademark of Excel Scientific, Inc.
Prevent evaporation
Thermostable adhesive
For PCR
RNase and DNase free
Chemically resistant
Transparent
Quickly seals microplates
Catalog No. 50-550-228 Supplier Research Products International Corp Supplier No. 202500
Adhesive films are specially designed for use as microplate covers. Prevent sample evaporation and contamination in sensitive PCR applications. RNase/DNase free. It is thermostable and functional from -40°C to +120°C at 75% humidity. ThermalSeal is a Trademark of Excel Scientific, Inc.
Prevent evaporation
Thermostable adhesive
For PCR
RNase and DNase free
Chemically resistant
Transparent
Quickly seals microplates