Learn More
Thomson Instrument Company Adhesive Foil Seal | Use w/ All 96 & 24 Well Plates
SDP
Catalog No. NC0268671
Encompass_Preferred
Thomson Instrument Company Adhesive Foil Seals are for short and long-term storage in well plates. Thomson well plates are offered in 24 and 96-well configurations with a V, U or pyramid shape bottom, and square or round wells for cell culture and analytical sample preparation. Well plates are designed to SLAS specifications and work well on automated liquid handling systems. Many plates are individually wrapped for your convince, and ideally designed to work with Thomson filter plates. Thomson manufactures its products in the USA to assure same day shipping.
Catalog No. NC0268671 Supplier Thomson Instrument Company Supplier No. 8994051
Thomson Instrument Company Adhesive Foil Seals are for short and long-term storage in well plates. Thomson well plates are offered in 24 and 96-well configurations with a V, U or pyramid shape bottom, and square or round wells for cell culture and analytical sample preparation. Well plates are designed to SLAS specifications and work well on automated liquid handling systems. Many plates are individually wrapped for your convince, and ideally designed to work with Thomson filter plates. Thomson manufactures its products in the USA to assure same day shipping.