Learn More
GA International Non-Adhesive Cryogenic Seal Tape (Patented) 1" x 10' / 25.4mm x 3m Red From -196°C to +260°C (-321°F to +500°F) / Minimum Application Temperature: -80°C (-112°F)
Catalog No. NC2817050
Encompass_Preferred
Non-adhesive, inert elastic cryogenic tape that bonds to itself to create a strong seal. Ideal for wrapping vials, bottles, cryo-boxes, plates, and other containers to be stored in deep-freeze and cryogenic conditions. The containers to be sealed can be sprayed with alcohol or other disinfecting solutions and then wrapped with CryoHUG™ to keep them sterile for extended periods of time and prevent contamination. This extraordinary tape can be put inside liquid nitrogen, freezers, and sterilized in autoclaves*, dry heat, and gamma irradiators. CryoHUG can be applied to frozen vials, and frozen containers freshly removed from -80°C freezers and liquid nitrogen tanks. Can be inscribed with permanent markers. Available in an assortment of colors as well as transparent.*In autoclaves, CryoHUG™ is not recommended for use on cap closed containers such as bottles, jars, and tubes.
Catalog No. NC2817050 Supplier GA International Supplier No. THG25C110RE
Non-adhesive, inert elastic cryogenic tape that bonds to itself to create a strong seal. Ideal for wrapping vials, bottles, cryo-boxes, plates, and other containers to be stored in deep-freeze and cryogenic conditions. The containers to be sealed can be sprayed with alcohol or other disinfecting solutions and then wrapped with CryoHUG™ to keep them sterile for extended periods of time and prevent contamination. This extraordinary tape can be put inside liquid nitrogen, freezers, and sterilized in autoclaves*, dry heat, and gamma irradiators. CryoHUG can be applied to frozen vials, and frozen containers freshly removed from -80°C freezers and liquid nitrogen tanks. Can be inscribed with permanent markers. Available in an assortment of colors as well as transparent.*In autoclaves, CryoHUG™ is not recommended for use on cap closed containers such as bottles, jars, and tubes.