Learn More
Ted Pella Inc Adhesive Pen for Paraffin Sections
SDP
Supplier:  Ted Pella Inc 22301
Encompass
Adhesive pen for paraffin sections. A coating of the glass slide will prevent paraffin sections from falling off or wrinkling. Will flatten sections in room temperature water. Stable to 120° C.
Catalog No. NC1265155