Learn More
Waters Corp Waters Adhesive Seal, 96 & 384-well Plates, Polyolefin Film, 100/pk
Catalog No. 50466287 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This adhesive seal features a polyolefin film with a synthetic rubber adhesive, specifically designed for protein and peptide sample preparation in buffers. Its adhesive is resistant to small percentages of acetonitrile and alcohol, and the film can be used across a temperature range of -80°C to 80°C without requiring heat sealing equipment.
  • Polyolefin film construction with a synthetic rubber adhesive.
  • Compatible with both 96-well and 384-well plates.
  • Usable across a temperature range of -80°C to 80°C.
  • Adhesive is resistant to small percentages of acetonitrile and alcohol in buffers.
  • Requires no heat sealing equipment for application.
  • Each pack contains 100 seals.
  • Ideal for sample preparation applications.
  • Compatible with automated liquid handling systems.
  • Designed to be easily peelable without leaving residue.
Catalog No. 50-466-287 Supplier Waters Corp Supplier No. 186006336
This adhesive seal features a polyolefin film with a synthetic rubber adhesive, specifically designed for protein and peptide sample preparation in buffers. Its adhesive is resistant to small percentages of acetonitrile and alcohol, and the film can be used across a temperature range of -80°C to 80°C without requiring heat sealing equipment.
  • Polyolefin film construction with a synthetic rubber adhesive.
  • Compatible with both 96-well and 384-well plates.
  • Usable across a temperature range of -80°C to 80°C.
  • Adhesive is resistant to small percentages of acetonitrile and alcohol in buffers.
  • Requires no heat sealing equipment for application.
  • Each pack contains 100 seals.
  • Ideal for sample preparation applications.
  • Compatible with automated liquid handling systems.
  • Designed to be easily peelable without leaving residue.