Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Adhesive Sealant

Catalog No. 19070913 Shop All 3M Products
Change view
Click to view available options
Quantity:
12 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-070-913 12 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-913 Supplier 3M™ Supplier No. 7000046609
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M Company Adhesive Sealant is a single-component, moisture-curing, elastomeric, isocyanate-free adhesive sealant that offers exceptional UV resistance and long term durability

  • Excellent UV resistance
  • Permanently elastic to allow joint movement
  • 10 to 30 minute skin time, paintable immediately
  • Silane Modified Polymer (SMP) adhesive sealant
  • Low VOC
  • NOT FOR RETAIL/CONSUMER USE
  • 3M Catalog# 60UV

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Panel to Frame/Stiffner to Panel Assembly
Color White
Model 60UV
Packaging Sausage pack
Quantity 12 Pk.
Volume (Metric) 600 ml
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.