Learn More
Agilent Technologies Adhesive Sealing Film, For 96-Well and 384-Well Plates, 100/Pk.
Catalog No. NC3104519
Encompass
This adhesive sealing film is designed for secure plate sealing during laboratory procedures. It is specifically compatible for use with 96-well and 384-well microplates.
  • Compatible with 96-well and 384-well plates
  • Supplied in a pack of 100
  • For research use only
Catalog No. NC3104519 Supplier Agilent Technologies Supplier No. 410186
This adhesive sealing film is designed for secure plate sealing during laboratory procedures. It is specifically compatible for use with 96-well and 384-well microplates.
  • Compatible with 96-well and 384-well plates
  • Supplied in a pack of 100
  • For research use only