Learn More
Bellco Glass, Inc. Adhesive sealing film for deep well plates, 100/pk
SDP
Catalog No. 502330576
Encompass_Preferred
Adhesive sealing film for deep well plates, 100/pk. adhesive sealing film minimizes evaporation in a wide range of temperatures in storage applications.
Catalog No. 50-233-0576 Supplier Bellco Glass, Inc. Supplier No. 451400632
Adhesive sealing film for deep well plates, 100/pk. adhesive sealing film minimizes evaporation in a wide range of temperatures in storage applications.