Learn More
Electron Microscopy Sciences Disposable Adhesive Slide Grids, 20 mm x 20 mm
SDP
Catalog No. 5034534
Encompass_Preferred
Adhesive grids stickers for microscope slides have side-by-side twin grids that are 20 mm x 20 mm each with 1 mm line spacing. They are used to Count or measure cells and estimate the size or length of microscopic or semi-micro objects.
They are compatible with conventional, inverted or dissecting microscopes.  Virtually all cells and specimens placed on top of Slide-Grids can be counted and/or sized. Use with dry specimens or suspended specimens.  Separate cover slips can be placed over each Slide-Grid.
Made of polyester with an acrylic adhesive. Slide-Grids are inert and stable under most laboratory conditions.
Catalog No. 50-345-34 Supplier Electron Microscopy Sciences Supplier No. 7036614
Adhesive grids stickers for microscope slides have side-by-side twin grids that are 20 mm x 20 mm each with 1 mm line spacing. They are used to Count or measure cells and estimate the size or length of microscopic or semi-micro objects.
They are compatible with conventional, inverted or dissecting microscopes.  Virtually all cells and specimens placed on top of Slide-Grids can be counted and/or sized. Use with dry specimens or suspended specimens.  Separate cover slips can be placed over each Slide-Grid.
Made of polyester with an acrylic adhesive. Slide-Grids are inert and stable under most laboratory conditions.