Learn More
Electron Microscopy Sciences CryoJane Adhesive Slides, 1x Coating 1
SDP
Catalog No. 5030412
Encompass
Glass slide with adhesive coating, when polymerized, bonds the still-frozen section from the tape to the slide, Avoid thawing the section, 1x coating slides for mild to normal tissues. For use with the CryoJane Tape-Transfer System.
Catalog No. 50-304-12 Supplier Electron Microscopy Sciences Supplier No. 628001X
Glass slide with adhesive coating, when polymerized, bonds the still-frozen section from the tape to the slide, Avoid thawing the section, 1x coating slides for mild to normal tissues. For use with the CryoJane Tape-Transfer System.