Learn More
Electron Microscopy Sciences CryoJane Adhesive Slides, 1/2x Coating 1
SDP
Catalog No. 5030404
Encompass
Glass slide with adhesive coating, when polymerized, bonds the still-frozen section from the tape to the slide, Avoid thawing the section, 1/2 x coating slides for mild to normal tissues. For use with the CryoJane Tape-Transfer System.
Catalog No. 50-304-04 Supplier Electron Microscopy Sciences Supplier No. 62800X
Glass slide with adhesive coating, when polymerized, bonds the still-frozen section from the tape to the slide, Avoid thawing the section, 1/2 x coating slides for mild to normal tissues. For use with the CryoJane Tape-Transfer System.