Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Adhesive Transfer Tape

Catalog No. 19072062 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
36 yd. x 3/4 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-072-062 36 yd. x 3/4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-062 Supplier 3M™ Supplier No. 7000047502
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Adhesive Transfer Tape offers instant adhesion to metals, glass, wood, papers and plastics.

  • Easy liner release for both manual and hand applications for high-speed flying spices and zero splices on most grades of paper in commercial print applications.
  • Bonds a wide variety of substrates including metal, glass, wood, paper, paint and many plastics
  • 60# Unbleached densified kraft liner allows for manual or hand applications
  • Provides consistently strong bonds in temperatures up to 250°F/121°C
  • Aggressive acrylic adhesive 400 bonds on contact
TRUSTED_SUSTAINABILITY

Specifications

Product Type Adhesive Transfer Tape
For Use With (Application) Zero-Speed and General Splicing
Adhesive Type Acrylic
Color Clear
Dimensions (L x W) 36 yd. x 3/4 in.
Material Acrylic
Model 465
Quantity 48 Rolls/Cs.
Thickness (English) 2 mil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.