Learn More
TARGETMOL CHEMICALS INC ADHP 10MG
SDP
Catalog No. 502581064
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ADHP (10-Acetyl-37-dihydroxyphenoxazine) is a fluorogenic peroxidase substrate ((lambda)ex=530 nm (lambda)em=590 nm). purity: 98%
Catalog No. 50-258-1064 Supplier TARGETMOL CHEMICALS INC Supplier No. T1891210MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ADHP (10-Acetyl-37-dihydroxyphenoxazine) is a fluorogenic peroxidase substrate ((lambda)ex=530 nm (lambda)em=590 nm). purity: 98%