Learn More
Cayman Chemical ADHP Fluorescent
SDP
Catalog No. 502886463
Encompass_Preferred
A highly sensitive, stable substrate for HRP that reacts with H2O2 to produce the fluorescent compound resorufin; enables detection of H2O2 at a concentration as low as 5 pmol per 100 µl samples
Catalog No. 50-288-6463 Supplier Cayman Chemical Supplier No. 100104691 MG
A highly sensitive, stable substrate for HRP that reacts with H2O2 to produce the fluorescent compound resorufin; enables detection of H2O2 at a concentration as low as 5 pmol per 100 µl samples