Learn More
Adipate/Phthalate, WS Proficiency Testing Material, MilliporeSigma™ Supelco™
Catalog No. 111017312 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
11-101-7312 2 mL
1 options
Catalog No. 11-101-7312 Supplier MilliporeSigma Supelco Supplier No. PE15962ML
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 mL
Concentration or Composition (by Analyte or Components) bis(2-Ethylhexyl) phthalate (DEHP) 5-50 μg/L; Butyl benzyl phthalate 10-50 μg/L; Di(2-ethylhexyl)adipate 8-50 μg/L; Di-n-butyl phthalate 10-50 μg/L; Di-n-octyl phthalate 10-50 μg/L; Diethyl phthalate 10-50 μg/L; Dimethyl phthalate 10-50 μg/L