Learn More
Apexbio Technology LLC Adiphenine HCl 50-42-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000088545
Encompass_Preferred
Adiphenine hydrochloride (CAS 50-42-0) is a small molecule that functions as an inhibitor of nicotinic acetylcholine receptors By antagonizing these receptors Adiphenine hydrochloride disrupts cholinergic neurotransmission at autonomic ganglia and neuromuscular junctions This activity underlies its utility as an antispasmodic agent where it modulates smooth muscle contraction in physiological and pharmacological studies Adiphenine hydrochloride is utilized in biomedical research to investigate nicotinic receptor signaling pathways and to explore mechanisms underlying smooth muscle relaxation
Catalog No. 50-000-88545 Supplier Apexbio Technology LLC Supplier No. B18785.1
Adiphenine hydrochloride (CAS 50-42-0) is a small molecule that functions as an inhibitor of nicotinic acetylcholine receptors By antagonizing these receptors Adiphenine hydrochloride disrupts cholinergic neurotransmission at autonomic ganglia and neuromuscular junctions This activity underlies its utility as an antispasmodic agent where it modulates smooth muscle contraction in physiological and pharmacological studies Adiphenine hydrochloride is utilized in biomedical research to investigate nicotinic receptor signaling pathways and to explore mechanisms underlying smooth muscle relaxation