Learn More
Medchemexpress LLC HY-B0379A 100mg Medchemexpress, Adiphenine (hydrochloride) CAS:50-42-0 Purity:>98%
SDP
Catalog No. 502082503
Encompass_Preferred
Medchemexpress, HY-B0379A 100mg Adiphenine (hydrochloride) CAS:50-42-0 Adiphenine hydrochloride is a non-competitive inhibitor of nicotinic acetylcholine receptor (nAChR), with an IC50s of 1.9, 1.8, 3.7, and 6.3 µM for α1, α3β4, α4β2, and α4β4, respectively. Adiphenine hydrochloride has narcotic and anticonvulsant effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2503 Supplier Medchemexpress LLC Supplier No. HYB0379A100MG
Medchemexpress, HY-B0379A 100mg Adiphenine (hydrochloride) CAS:50-42-0 Adiphenine hydrochloride is a non-competitive inhibitor of nicotinic acetylcholine receptor (nAChR), with an IC50s of 1.9, 1.8, 3.7, and 6.3 µM for α1, α3β4, α4β2, and α4β4, respectively. Adiphenine hydrochloride has narcotic and anticonvulsant effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.