Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adiphenine hydrochloride | 50-42-0 | 99.1% | 500 MG
SDP

Catalog No. 5000406106
Encompass_Preferred

Adiphenine hydrochloride is a non-competitive inhibitor of the nicotinic acetylcholine receptor (nAChR). It exhibits anticonvulsant effects and has specific IC50 values for various nAChR subtypes including α1, α3β4, α4β2, and α4β4. In vivo, it prevents hindleg tonic-extensor components of maximal electroshock seizures (MES) in mice.

  • Non-competitive inhibitor of nicotinic acetylcholine receptor (nAChR)
  • Exhibits anticonvulsant effects
  • Specific IC50 values for α1, α3β4, α4β2, and α4β4 nAChR subtypes
  • Prevents hindleg tonic-extensor components of maximal electroshock seizures (MES) in mice

Catalog No. 50-004-06106 Supplier Medchemexpress LLC Supplier No. HYB0379A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adiphenine hydrochloride is a non-competitive inhibitor of the nicotinic acetylcholine receptor (nAChR). It exhibits anticonvulsant effects and has specific IC50 values for various nAChR subtypes including α1, α3β4, α4β2, and α4β4. In vivo, it prevents hindleg tonic-extensor components of maximal electroshock seizures (MES) in mice.

  • Non-competitive inhibitor of nicotinic acetylcholine receptor (nAChR)
  • Exhibits anticonvulsant effects
  • Specific IC50 values for α1, α3β4, α4β2, and α4β4 nAChR subtypes
  • Prevents hindleg tonic-extensor components of maximal electroshock seizures (MES) in mice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.