Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adiphenine hydrochloride | 50-42-0 | MFCD00008899 | 99.1% | 1 ML
SDP

Catalog No. 5000406404
Encompass_Preferred

Adiphenine hydrochloride is a non-competitive inhibitor of nicotinic acetylcholine receptor (nAChR), exhibiting specific IC50s for various receptor subtypes. This compound has also demonstrated anticonvulsant effects in studies.

  • Non-competitive inhibitor of nicotinic acetylcholine receptor (nAChR)
  • Exhibits anticonvulsant effects
  • Blocks function of α1*-nAChR, α3α4*-nAChR, α4β2- and α4β4-nAChR in various cell lines
  • Decreases frequency of ACh-induced single-channel currents
  • Prevents hindleg tonic-extensor component of maximal electroshock seizures in mice

Catalog No. 50-004-06404 Supplier Medchemexpress LLC Supplier No. HYB0379A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adiphenine hydrochloride is a non-competitive inhibitor of nicotinic acetylcholine receptor (nAChR), exhibiting specific IC50s for various receptor subtypes. This compound has also demonstrated anticonvulsant effects in studies.

  • Non-competitive inhibitor of nicotinic acetylcholine receptor (nAChR)
  • Exhibits anticonvulsant effects
  • Blocks function of α1*-nAChR, α3α4*-nAChR, α4β2- and α4β4-nAChR in various cell lines
  • Decreases frequency of ACh-induced single-channel currents
  • Prevents hindleg tonic-extensor component of maximal electroshock seizures in mice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.