Learn More
Medchemexpress LLC Adipic acid | 124-04-9 | MFCD00004420 | 99.3% | 146.14 g/mol | C6H10O4 | 500 MG
SDP
Catalog No. 5000299979
Encompass_Preferred
Adipic acid (hexanedioic acid) analytical standard intended for research and analytical use. This white crystalline dicarboxylic acid is commonly used as a reference material in chromatographic and spectrometric methods, and as a reagent in synthetic and formulation studies.
  • Analytical standard suitable for method validation and QC.
  • High purity for reliable and reproducible results.
  • Known molecular weight and formula for accurate calculations.
  • Compatible with chromatographic and spectrometric analyses.
  • Available in small laboratory pack sizes for convenient use.
Catalog No. 50-002-99979 Supplier Medchemexpress LLC Supplier No. HYW017522R500MG
Adipic acid (hexanedioic acid) analytical standard intended for research and analytical use. This white crystalline dicarboxylic acid is commonly used as a reference material in chromatographic and spectrometric methods, and as a reagent in synthetic and formulation studies.
  • Analytical standard suitable for method validation and QC.
  • High purity for reliable and reproducible results.
  • Known molecular weight and formula for accurate calculations.
  • Compatible with chromatographic and spectrometric analyses.
  • Available in small laboratory pack sizes for convenient use.