Learn More
Medchemexpress LLC Adipic acid | 124-04-9 | MFCD00004420 | 99.8% | 146.14 g/mol | C6H10O4 | 1 ML
SDP
Catalog No. 5000301447
Encompass_Preferred
Adipic acid (hexanedioic acid) is a small dicarboxylic organic acid used as a reagent, analytical standard, and building block in chemical synthesis. It is reported to have low toxicity and has shown oral activity with anti-HSV-1 effects in some studies. Typical applications include analytical reference standards, metabolic research, and polymer and resin synthesis.
  • Analytical standard suitable for research applications.
  • High purity for accurate analytical results.
  • Stable white crystalline solid with a defined molecular weight.
  • Available in solution formats for assay convenience.
  • Used in synthesis of polymers, resins, and industrial chemicals.
Catalog No. 50-003-01447 Supplier Medchemexpress LLC Supplier No. HYW01752210MM/1ML
Adipic acid (hexanedioic acid) is a small dicarboxylic organic acid used as a reagent, analytical standard, and building block in chemical synthesis. It is reported to have low toxicity and has shown oral activity with anti-HSV-1 effects in some studies. Typical applications include analytical reference standards, metabolic research, and polymer and resin synthesis.
  • Analytical standard suitable for research applications.
  • High purity for accurate analytical results.
  • Stable white crystalline solid with a defined molecular weight.
  • Available in solution formats for assay convenience.
  • Used in synthesis of polymers, resins, and industrial chemicals.