Learn More
Medchemexpress LLC Adipic acid | 124-04-9 | MFCD00004420 | 99.8% | 146.14 g/mol | C6H10O4 | 100 G
SDP
Catalog No. 5000300376
Encompass_Preferred
Adipic acid (hexanedioic acid, CAS 124-04-9) is a white crystalline dicarboxylic acid provided as a high-purity research reagent. It serves as a versatile synthetic building block and analytical standard with applications in polymer synthesis, materials research, and food additive studies; it has low toxicity and reported antiviral activity.
  • High purity (99.82%) suitable for analytical use.
  • Molecular weight 146.14 g/mol.
  • Useful as a building block for polymer and resin research.
  • Applicable as an analytical standard and reagent in labs.
  • Low toxicity and reported anti-HSV-1 activity.
  • Available in 100 g pack size for lab use.
Catalog No. 50-003-00376 Supplier Medchemexpress LLC Supplier No. HYW017522100G
Adipic acid (hexanedioic acid, CAS 124-04-9) is a white crystalline dicarboxylic acid provided as a high-purity research reagent. It serves as a versatile synthetic building block and analytical standard with applications in polymer synthesis, materials research, and food additive studies; it has low toxicity and reported antiviral activity.
  • High purity (99.82%) suitable for analytical use.
  • Molecular weight 146.14 g/mol.
  • Useful as a building block for polymer and resin research.
  • Applicable as an analytical standard and reagent in labs.
  • Low toxicity and reported anti-HSV-1 activity.
  • Available in 100 g pack size for lab use.