Learn More
Medchemexpress LLC Adipic acid-13C | 2708283-72-9 | 98.6% | 147.13 | C513CH10O4 | 25 MG
SDP
Catalog No. 5000303601
Encompass_Preferred
Adipic acid-13C is a 13C-stable-isotope labeled adipic (hexanedioic) acid supplied as a white to off-white solid for use as an internal standard, tracer, or reference in analytical, metabolic, and mass spectrometry applications requiring isotopically labeled adipic acid.
  • Isotopically labeled for use as an internal standard in analytical assays.
  • High purity (>98%) suitable for quantitative studies.
  • Supplied as a white to off-white solid for easy handling.
  • Multiple pack sizes available for small-scale experiments.
  • Soluble in DMSO with manufacturer-recommended stock solutions.
  • Stable when stored under recommended conditions.
Catalog No. 50-003-03601 Supplier Medchemexpress LLC Supplier No. 25MGHYW017522S3
Adipic acid-13C is a 13C-stable-isotope labeled adipic (hexanedioic) acid supplied as a white to off-white solid for use as an internal standard, tracer, or reference in analytical, metabolic, and mass spectrometry applications requiring isotopically labeled adipic acid.
  • Isotopically labeled for use as an internal standard in analytical assays.
  • High purity (>98%) suitable for quantitative studies.
  • Supplied as a white to off-white solid for easy handling.
  • Multiple pack sizes available for small-scale experiments.
  • Soluble in DMSO with manufacturer-recommended stock solutions.
  • Stable when stored under recommended conditions.