Learn More
Medchemexpress LLC Adipic acid-13C | 2708283-72-9 | 98.6% | 147.13 g/mol | C5(13C)3CH10O4 | 100 MG
SDP
Catalog No. 5000300381
Encompass_Preferred
Adipic acid-13C is the 13C isotopically labeled form of adipic (hexanedioic) acid intended for analytical and research applications. It serves as a stable-isotope reference standard that retains the chemical properties of unlabeled adipic acid while enabling isotopic tracing and quantitative analysis in mass spectrometry and metabolic studies.
  • Stable-isotope labeled for mass spectrometry and tracing studies.
  • Suitable as an internal standard for quantitative analytical workflows.
  • Reported purity 98.62% as supplied.
  • Provided in small, laboratory-scale quantities for research use.
  • Soluble in common organic solvents such as DMSO for sample preparation.
Catalog No. 50-003-00381 Supplier Medchemexpress LLC Supplier No. HYW017522S3100MG
Adipic acid-13C is the 13C isotopically labeled form of adipic (hexanedioic) acid intended for analytical and research applications. It serves as a stable-isotope reference standard that retains the chemical properties of unlabeled adipic acid while enabling isotopic tracing and quantitative analysis in mass spectrometry and metabolic studies.
  • Stable-isotope labeled for mass spectrometry and tracing studies.
  • Suitable as an internal standard for quantitative analytical workflows.
  • Reported purity 98.62% as supplied.
  • Provided in small, laboratory-scale quantities for research use.
  • Soluble in common organic solvents such as DMSO for sample preparation.