Learn More
Medchemexpress LLC Adipic acid-13C | 2708283-72-9 | 98.6% | 147.13 g/mol | C5(13C)H10O4 | 5 MG
SDP
Catalog No. 5000300425
Encompass_Preferred
Adipic acid-13C is the 13C-isotope labeled form of adipic (hexanedioic) acid for research and analytical use as a stable isotope reference standard. It is a white to off-white solid suitable for mass spectrometry, NMR, and tracer studies; typical purity is 98.62% and molecular weight is 147.13 g/mol.
  • 13C-labeled for use as an internal standard in MS and NMR analyses.
  • High purity (98.62%) for accurate quantification and reproducible results.
  • Solid, easy to handle and store under recommended conditions.
  • Available in small pack sizes (including 5 mg) for analytical applications.
  • Suitable for tracer and metabolite identification studies.
Catalog No. 50-003-00425 Supplier Medchemexpress LLC Supplier No. HYW017522S35MG
Adipic acid-13C is the 13C-isotope labeled form of adipic (hexanedioic) acid for research and analytical use as a stable isotope reference standard. It is a white to off-white solid suitable for mass spectrometry, NMR, and tracer studies; typical purity is 98.62% and molecular weight is 147.13 g/mol.
  • 13C-labeled for use as an internal standard in MS and NMR analyses.
  • High purity (98.62%) for accurate quantification and reproducible results.
  • Solid, easy to handle and store under recommended conditions.
  • Available in small pack sizes (including 5 mg) for analytical applications.
  • Suitable for tracer and metabolite identification studies.