Learn More
Medchemexpress LLC Adipic acid-13C | 2708283-72-9 | 98.6% | 152.10 g/mol | 13C6H10O4 | 50 MG
SDP
Catalog No. 5000301786
Encompass_Preferred
Adipic acid-13C is the carbon-13 isotopically labeled form of adipic (hexanedioic) acid supplied as a stable isotope standard for analytical and research use. It is intended for metabolic tracing, mass spectrometry internal standards, and other applications that require an isotopically labeled compound; the material is provided as a solid with high isotopic enrichment and chemical purity.
  • Stable isotope-labeled analog for tracer studies.
  • High isotopic enrichment (13C6) suitable for mass spectrometry.
  • High chemical purity for analytical applications.
  • Provided as a solid for convenient storage and handling.
  • Suitable as an internal standard for quantitative assays.
Catalog No. 50-003-01786 Supplier Medchemexpress LLC Supplier No. HYW017522S350MG
Adipic acid-13C is the carbon-13 isotopically labeled form of adipic (hexanedioic) acid supplied as a stable isotope standard for analytical and research use. It is intended for metabolic tracing, mass spectrometry internal standards, and other applications that require an isotopically labeled compound; the material is provided as a solid with high isotopic enrichment and chemical purity.
  • Stable isotope-labeled analog for tracer studies.
  • High isotopic enrichment (13C6) suitable for mass spectrometry.
  • High chemical purity for analytical applications.
  • Provided as a solid for convenient storage and handling.
  • Suitable as an internal standard for quantitative assays.