Learn More
Medchemexpress LLC Adipic acid-13C6 | 942037-55-0 | 98.0% | 152.10 g/mol | 13C6H10O4 | 1 MG
SDP
Catalog No. 5000300416
Encompass_Preferred
Adipic acid-13C6 is the 13C6-isotopically labeled form of adipic acid used as an internal standard and tracer in analytical chemistry and metabolic studies. Supplied as a stable, high-purity powder for research use only, it enables precise quantification in mass spectrometry and isotope dilution assays.
  • Isotopically labeled 13C6 standard for precise mass spectrometry quantification.
  • High purity suitable for analytical and quantitative workflows.
  • Stable powder form for reliable long-term storage and handling.
  • Widely used as an internal standard and tracer in metabolic studies.
  • Suitable for isotope dilution assays and quantitative LC-MS workflows.
Catalog No. 50-003-00416 Supplier Medchemexpress LLC Supplier No. HYW017522S11MG
Adipic acid-13C6 is the 13C6-isotopically labeled form of adipic acid used as an internal standard and tracer in analytical chemistry and metabolic studies. Supplied as a stable, high-purity powder for research use only, it enables precise quantification in mass spectrometry and isotope dilution assays.
  • Isotopically labeled 13C6 standard for precise mass spectrometry quantification.
  • High purity suitable for analytical and quantitative workflows.
  • Stable powder form for reliable long-term storage and handling.
  • Widely used as an internal standard and tracer in metabolic studies.
  • Suitable for isotope dilution assays and quantitative LC-MS workflows.