Learn More
Medchemexpress LLC Adipic acid-13C6 | 942037-55-0 | 98.0% | 152.10 g/mol | 13C6H10O4 | 5 MG
SDP
Catalog No. 5000299429
Encompass_Preferred
Adipic acid-13C6 is the 13C-labeled isotopologue of adipic (hexanedioic) acid used as an internal standard, tracer, and reference standard in mass spectrometry and metabolic studies. It provides defined isotopic enrichment for quantitative analysis and isotope tracing experiments.
  • Isotopically labeled internal standard for mass spectrometry.
  • High isotopic enrichment suitable for quantitative assays.
  • Stable, non-radioactive tracer for metabolic pathway studies.
  • Available in small, pre-weighed quantities for analytical workflows.
  • Compatible with LC-MS and GC-MS sample preparation.
Catalog No. 50-002-99429 Supplier Medchemexpress LLC Supplier No. HYW017522S15MG
Adipic acid-13C6 is the 13C-labeled isotopologue of adipic (hexanedioic) acid used as an internal standard, tracer, and reference standard in mass spectrometry and metabolic studies. It provides defined isotopic enrichment for quantitative analysis and isotope tracing experiments.
  • Isotopically labeled internal standard for mass spectrometry.
  • High isotopic enrichment suitable for quantitative assays.
  • Stable, non-radioactive tracer for metabolic pathway studies.
  • Available in small, pre-weighed quantities for analytical workflows.
  • Compatible with LC-MS and GC-MS sample preparation.