Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adipic acid-13C6 (hexanedioic acid-13C6) | 942037-55-0 | 98.0% | 152.10 g·mol⁻1 | 13C6H10O4 | 10 MG
SDP

Catalog No. 5000301384
Encompass_Preferred

Adipic acid-13C6 is the six-carbon 13C-labeled isotopologue of adipic (hexanedioic) acid, provided as a powder for analytical and research use. It is intended as a stable isotope reference standard for mass spectrometry, metabolic tracing, and isotopic labeling experiments.

  • Stable 13C6 labeling suitable for mass spectrometry internal standard applications.
  • High chemical purity of 98.0% to support reliable analytical results.
  • Small laboratory sizes available, including 10 mg, for trace-level experiments.
  • Powder form that is easy to weigh and dissolve for solution preparation.
  • Recommended storage at -20°C (powder) to preserve stability over time.
  • Molecular weight 152.10 g·mol⁻1 and formula 13C6H10O4 for reference.

Catalog No. 50-003-01384 Supplier Medchemexpress LLC Supplier No. HYW017522S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adipic acid-13C6 is the six-carbon 13C-labeled isotopologue of adipic (hexanedioic) acid, provided as a powder for analytical and research use. It is intended as a stable isotope reference standard for mass spectrometry, metabolic tracing, and isotopic labeling experiments.

  • Stable 13C6 labeling suitable for mass spectrometry internal standard applications.
  • High chemical purity of 98.0% to support reliable analytical results.
  • Small laboratory sizes available, including 10 mg, for trace-level experiments.
  • Powder form that is easy to weigh and dissolve for solution preparation.
  • Recommended storage at -20°C (powder) to preserve stability over time.
  • Molecular weight 152.10 g·mol⁻1 and formula 13C6H10O4 for reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.