Learn More
Medchemexpress LLC Adipic acid-d10 | 25373-21-1 | MFCD00190459 | 99.9% | 156.20 | C6D10O4 | 50 MG
SDP
Catalog No. 5000300188
Encompass_Preferred
Adipic acid-d10 is the fully deuterated isotopologue of adipic acid used as a stable-isotope analytical standard and tracer for method development. It provides high isotopic enrichment and chemical purity for reliable quantitation in research-scale analytical workflows.
  • Deuterated stable-isotope standard for analytical applications.
  • High isotopic enrichment and chemical purity (reported 99.93%).
  • Suitable as an internal standard for mass spectrometry and NMR.
  • Formula C6D10O4 and molecular weight 156.20 for accurate calculations.
  • Available in small research-scale quantities for method development.
Catalog No. 50-003-00188 Supplier Medchemexpress LLC Supplier No. HYW017522S50MG
Add to Cart
Add to Cart
Adipic acid-d10 is the fully deuterated isotopologue of adipic acid used as a stable-isotope analytical standard and tracer for method development. It provides high isotopic enrichment and chemical purity for reliable quantitation in research-scale analytical workflows.
  • Deuterated stable-isotope standard for analytical applications.
  • High isotopic enrichment and chemical purity (reported 99.93%).
  • Suitable as an internal standard for mass spectrometry and NMR.
  • Formula C6D10O4 and molecular weight 156.20 for accurate calculations.
  • Available in small research-scale quantities for method development.