Learn More
Medchemexpress LLC Adipic acid-d10 | 25373-21-1 | MFCD00190459 | 99.9% | 156.20 g·mol⁻¹ | C6D10O4 | 100 MG
SDP
Catalog No. 5000300414
Encompass_Preferred
Adipic acid-d10 is the deuterium-labeled analogue of adipic acid (C6D10O4, MW 156.20 g·mol⁻¹; CAS 25373-21-1) formulated for use as a tracer and internal standard in quantitative analytical workflows. It provides consistent isotopic enrichment and high chemical purity to support accurate identification and reliable quantitation in NMR, GC-MS, and LC-MS applications.
  • High chemical purity suitable for analytical applications.
  • Consistent isotopic labeling for reliable internal standard performance.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Available in small package sizes for laboratory use.
  • Facilitates accurate quantitation and peak identification.
  • Stable under typical storage conditions for labeled standards.
Catalog No. 50-003-00414 Supplier Medchemexpress LLC Supplier No. HYW017522S100MG
Adipic acid-d10 is the deuterium-labeled analogue of adipic acid (C6D10O4, MW 156.20 g·mol⁻¹; CAS 25373-21-1) formulated for use as a tracer and internal standard in quantitative analytical workflows. It provides consistent isotopic enrichment and high chemical purity to support accurate identification and reliable quantitation in NMR, GC-MS, and LC-MS applications.
  • High chemical purity suitable for analytical applications.
  • Consistent isotopic labeling for reliable internal standard performance.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Available in small package sizes for laboratory use.
  • Facilitates accurate quantitation and peak identification.
  • Stable under typical storage conditions for labeled standards.