Learn More
Medchemexpress LLC Adipic acid-d10 | 25373-21-1 | MFCD00190459 | 99.9% | 156.20 g/mol | C6D10O4 | 25 MG
SDP
Catalog No. 5000303257
Encompass_Preferred
Adipic acid-d10 is the deuterium-labeled isotopologue of adipic acid supplied for research use as a stable-isotope tracer and internal standard for quantitative analysis. It provides high chemical purity and deuterium enrichment suitable for NMR, GC-MS, and LC-MS method development and isotopic labeling studies.
  • Used as a stable-isotope tracer for quantitative studies.
  • Serves as an internal standard for NMR, GC-MS, and LC-MS.
  • High chemical purity and deuterium enrichment for reliable results.
  • Suitable for analytical method development and validation.
  • Supplied for research use only; not for human consumption.
Catalog No. 50-003-03257 Supplier Medchemexpress LLC Supplier No. 25MGHYW017522S
Adipic acid-d10 is the deuterium-labeled isotopologue of adipic acid supplied for research use as a stable-isotope tracer and internal standard for quantitative analysis. It provides high chemical purity and deuterium enrichment suitable for NMR, GC-MS, and LC-MS method development and isotopic labeling studies.
  • Used as a stable-isotope tracer for quantitative studies.
  • Serves as an internal standard for NMR, GC-MS, and LC-MS.
  • High chemical purity and deuterium enrichment for reliable results.
  • Suitable for analytical method development and validation.
  • Supplied for research use only; not for human consumption.