Learn More
Medchemexpress LLC Adipic acid-d10 | 25373-21-1 | MFCD00190459 | 99.9% | 156.20 g·mol⁻¹ | C6D10O4 | 5 MG
SDP
Catalog No. 5000300496
Encompass_Preferred
Adipic acid-d10 is the deuterated isotopologue of adipic acid supplied as a high-purity stable isotope standard for analytical and research use. It is formulated for accurate mass-shift identification in mass spectrometry and as an internal standard for NMR and isotopic tracing studies.
  • High purity (99.93%) suitable for analytical standards.
  • Deuterated chemical formula C6D10O4 for clear mass shifts in mass spectrometry.
  • Suitable as an internal standard for mass spectrometry and NMR.
  • Useful for isotopic tracing and metabolic or synthetic studies.
  • Available in small, precisely weighed packages for accurate dosing.
Catalog No. 50-003-00496 Supplier Medchemexpress LLC Supplier No. HYW017522S5MG
Adipic acid-d10 is the deuterated isotopologue of adipic acid supplied as a high-purity stable isotope standard for analytical and research use. It is formulated for accurate mass-shift identification in mass spectrometry and as an internal standard for NMR and isotopic tracing studies.
  • High purity (99.93%) suitable for analytical standards.
  • Deuterated chemical formula C6D10O4 for clear mass shifts in mass spectrometry.
  • Suitable as an internal standard for mass spectrometry and NMR.
  • Useful for isotopic tracing and metabolic or synthetic studies.
  • Available in small, precisely weighed packages for accurate dosing.