Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adipic acid-d4 | 121311-78-2 | 98.0% | 150.17 g/mol | C6H6D4O4 | 50 MG
SDP

Catalog No. 5000299985
Encompass_Preferred

Adipic acid-d4 is the deuterium-labeled form of adipic (hexanedioic) acid used as an internal standard and reference material in analytical, metabolic, and isotope-tracing studies. Supplied as a high-purity, stable powder, it is suited for quantitative mass spectrometry and chromatographic workflows where isotopic labeling improves accuracy and peak identification.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity suitable for analytical use.
  • Molecular formula C6H6D4O4; molecular weight 150.17 g/mol.
  • Available in small research pack sizes for method development.
  • Stable powder form with recommended cold storage to preserve integrity.
  • Compatible with mass spectrometry and chromatographic methods.

Catalog No. 50-002-99985 Supplier Medchemexpress LLC Supplier No. HYW017522S250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adipic acid-d4 is the deuterium-labeled form of adipic (hexanedioic) acid used as an internal standard and reference material in analytical, metabolic, and isotope-tracing studies. Supplied as a high-purity, stable powder, it is suited for quantitative mass spectrometry and chromatographic workflows where isotopic labeling improves accuracy and peak identification.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity suitable for analytical use.
  • Molecular formula C6H6D4O4; molecular weight 150.17 g/mol.
  • Available in small research pack sizes for method development.
  • Stable powder form with recommended cold storage to preserve integrity.
  • Compatible with mass spectrometry and chromatographic methods.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.