Learn More
Medchemexpress LLC Adipic acid-d4 | 121311-78-2 | 98.0% | 150.170 g/mol | C6H6D4O4 | 10 MG
SDP
Catalog No. 5000299984
Encompass_Preferred
Adipic acid-d4 is a deuterium-labeled stable isotope of adipic acid (CAS 121311-78-2) supplied as a white to off-white solid at 98.0% purity. It is intended for use as an internal standard, tracer, or analytical reference in mass spectrometry and isotopic studies, and is provided in small research pack sizes suitable for quantitative analysis. Store powder at -20°C (3 years) or 4°C (2 years); in solvent store at -80°C (6 months).
  • Deuterium-labeled internal standard for mass spectrometry and isotope tracing.
  • High purity (98.0%) for reliable analytical performance.
  • White to off-white solid suitable for preparation of standard solutions.
  • Available in small pack sizes for analytical and research use.
  • Suitable for quantitative analysis, method development, and tracer studies.
  • Recommended storage conditions provided to preserve stability.
Catalog No. 50-002-99984 Supplier Medchemexpress LLC Supplier No. HYW017522S210MG
Adipic acid-d4 is a deuterium-labeled stable isotope of adipic acid (CAS 121311-78-2) supplied as a white to off-white solid at 98.0% purity. It is intended for use as an internal standard, tracer, or analytical reference in mass spectrometry and isotopic studies, and is provided in small research pack sizes suitable for quantitative analysis. Store powder at -20°C (3 years) or 4°C (2 years); in solvent store at -80°C (6 months).
  • Deuterium-labeled internal standard for mass spectrometry and isotope tracing.
  • High purity (98.0%) for reliable analytical performance.
  • White to off-white solid suitable for preparation of standard solutions.
  • Available in small pack sizes for analytical and research use.
  • Suitable for quantitative analysis, method development, and tracer studies.
  • Recommended storage conditions provided to preserve stability.