Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adipic acid-d4 | 121311-78-2 | MFCD01355072 | 98.0% | 150.17 g/mol | C6H6D4O4 | 100 MG
SDP

Catalog No. 5000299444
Encompass_Preferred

Adipic acid-d4 is the deuterated, stable-isotope form of adipic (hexanedioic) acid used as a reference standard and tracer in analytical, metabolic, and research applications.

  • Purity 98.0%.
  • Formula C6H6D4O4.
  • Molecular weight 150.17 g/mol.
  • Intended for use as a stable-isotope reference standard in quantitative and qualitative analyses.
  • Available in reference package sizes including 5 mg, 10 mg, 25 mg, 50 mg, and 100 mg.

Catalog No. 50-002-99444 Supplier Medchemexpress LLC Supplier No. HYW017522S2100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adipic acid-d4 is the deuterated, stable-isotope form of adipic (hexanedioic) acid used as a reference standard and tracer in analytical, metabolic, and research applications.

  • Purity 98.0%.
  • Formula C6H6D4O4.
  • Molecular weight 150.17 g/mol.
  • Intended for use as a stable-isotope reference standard in quantitative and qualitative analyses.
  • Available in reference package sizes including 5 mg, 10 mg, 25 mg, 50 mg, and 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.