Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adipic acid-d4 (hexanedioic acid-d4) | 121311-78-2 | 98.0% | 150.17 g/mol | C6H6D4O4 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYW017522S25MG

Encompass_Preferred

Adipic acid-d4 is a deuterium-labeled stable isotope of adipic (hexanedioic) acid supplied as a small, pre-weighed research reagent and analytical standard for use in mass spectrometry, tracer studies, and methods requiring isotopically labeled compounds.

  • Deuterium-labeled stable isotope for analytical applications.
  • High purity (98.0%) suitable for quantitative analysis.
  • Available in small pre-weighed packages including 5 mg.
  • Good solubility in DMSO (≥100 mg/mL) for solution preparation.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years).
  • Molecular formula C6H6D4O4; molecular weight 150.17 g/mol.

Catalog No. 50-002-99432


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.