Learn More
Medchemexpress LLC Adipic acid-d8 | 52089-65-3 | 99.7% | 154.19 g/mol | C6H2D8O4 | 100 MG
SDP
Catalog No. 5000300497
Encompass_Preferred
Adipic acid-d8 is the deuterium-labeled form of adipic (hexanedioic) acid supplied as a high-purity isotopic standard for analytical and research applications. It is used as an internal standard, tracer, or calibration reference in mass spectrometry, NMR, and method development.
  • Deuterium-labeled (d8) for clear mass shifts in MS analyses.
  • High purity suitable for use as an analytical reference material.
  • Compact 100 mg quantity convenient for laboratory workflows.
  • Known molecular weight and formula for accurate calculations.
  • Suitable for quantitative LC-MS/MS, GC-MS, and NMR applications.
Catalog No. 50-003-00497 Supplier Medchemexpress LLC Supplier No. HYW017522S4100MG
Adipic acid-d8 is the deuterium-labeled form of adipic (hexanedioic) acid supplied as a high-purity isotopic standard for analytical and research applications. It is used as an internal standard, tracer, or calibration reference in mass spectrometry, NMR, and method development.
  • Deuterium-labeled (d8) for clear mass shifts in MS analyses.
  • High purity suitable for use as an analytical reference material.
  • Compact 100 mg quantity convenient for laboratory workflows.
  • Known molecular weight and formula for accurate calculations.
  • Suitable for quantitative LC-MS/MS, GC-MS, and NMR applications.