Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adipic acid-d8 | 52089-65-3 | 99.7% | 154.19 g/mol | C6H2D8O4 | 25 MG
SDP

Catalog No. 5000303188
Encompass_Preferred

Adipic acid-d8 (hexanedioic acid-d8) is a deuterium-labeled adipic acid used as a stable isotope standard and tracer in analytical chemistry. Supplied as a high-purity solid, it supports accurate quantitation and method development in mass spectrometry and related analytical workflows.

  • High isotopic enrichment and chemical purity for accurate mass spectrometry.
  • Solid form simplifies storage and handling.
  • Available in small milligram pack sizes for analytical workflows.
  • Molecular formula C6H2D8O4; molecular weight 154.19 g/mol.
  • Commonly used as an internal standard, tracer, and analytical reference.

Catalog No. 50-003-03188 Supplier Medchemexpress LLC Supplier No. 25MGHYW017522S4
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Adipic acid-d8 (hexanedioic acid-d8) is a deuterium-labeled adipic acid used as a stable isotope standard and tracer in analytical chemistry. Supplied as a high-purity solid, it supports accurate quantitation and method development in mass spectrometry and related analytical workflows.

  • High isotopic enrichment and chemical purity for accurate mass spectrometry.
  • Solid form simplifies storage and handling.
  • Available in small milligram pack sizes for analytical workflows.
  • Molecular formula C6H2D8O4; molecular weight 154.19 g/mol.
  • Commonly used as an internal standard, tracer, and analytical reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.