Learn More
Medchemexpress LLC Adipic acid dihydrazide-d8 | 1202865-37-9 | 99.0% | 10 MG
SDP
Catalog No. 5000297098
Encompass_Preferred
Adipic acid dihydrazide-d8 is the deuterium-labeled isotopologue of adipic acid dihydrazide intended for research use. It is used as a stable isotope standard or tracer in analytical and synthetic workflows, particularly mass spectrometry and quantitative method development. The compound is supplied in small quantities for reference and is stored as a powder at low temperature to maintain stability.
  • Deuterium-labeled internal standard for mass spectrometry and quantitation.
  • Reported purity 99% suitable for analytical applications.
  • Supplied in small vial sizes for use as reference standards or tracers.
  • Powder is stable at -20°C for long-term storage.
  • Applicable to method development, metabolite tracing, and analytical validation.
Catalog No. 50-002-97098 Supplier Medchemexpress LLC Supplier No. HYD0200S10MG
Adipic acid dihydrazide-d8 is the deuterium-labeled isotopologue of adipic acid dihydrazide intended for research use. It is used as a stable isotope standard or tracer in analytical and synthetic workflows, particularly mass spectrometry and quantitative method development. The compound is supplied in small quantities for reference and is stored as a powder at low temperature to maintain stability.
  • Deuterium-labeled internal standard for mass spectrometry and quantitation.
  • Reported purity 99% suitable for analytical applications.
  • Supplied in small vial sizes for use as reference standards or tracers.
  • Powder is stable at -20°C for long-term storage.
  • Applicable to method development, metabolite tracing, and analytical validation.