Learn More
Medchemexpress LLC Adipic acid dihydrazide-d8 | 1202865-37-9 | 99% | 5 MG
SDP
Catalog No. 5000411047
Encompass_Preferred
Adipic acid dihydrazide-d8 is the deuterium labeled Adipic acid dihydrazide. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-11047 Supplier Medchemexpress LLC Supplier No. HYD0200S5MG
Adipic acid dihydrazide-d8 is the deuterium labeled Adipic acid dihydrazide. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.