Learn More
ALADDIN SCIENTIFIC CORPORATION Adipic acid dihydrazideADH, 100g
Supplier:  ALADDIN SCIENTIFIC CORPORATION A109760100G
Encompass_Preferred
Homobifunctional cross-linking reagent that is specific for aldehydes resulting in relatively stable hydrazone linkages This is commonly used in the linking of glycoproteins such as antibodies in a site specific fashion following periodate oxidation Oxidation and coupling may conveniently be performed at pH 5 0 due to the low pKa of the hydrazide which avoids competition by primary amines Note that the hydrazone bond may be stabilized by selective reduction using for example sodium cyanoborohydride General descriptionAdipic acid dihydrazide is a homobifunctional cross-linking reagent that is specific for aldehydes This results in relatively stable hydrazone linkages Adipic acid dihydrazide is generally used in the linking of glycoproteins like antibodies in a site-specific fashion following periodate oxidation Oxidation and coupling may be performed at pH 5 0 due to the low pKa of the hydrazide which avoids competition by primary amines
Catalog No. 50-000-82510