Learn More
Medchemexpress LLC Adipic acid (hexanedioic acid) | 124-04-9 | MFCD00004420 | 99.8% | 146.14 g/mol | C6H10O4 | 500 G
SDP
Catalog No. 5000301783
Encompass_Preferred
Adipic acid (hexanedioic acid) is a straight-chain dicarboxylic acid supplied as a crystalline solid for laboratory use. It is commonly used as a reagent, chemical intermediate, and analytical standard; manufacturer listings report CAS 124-04-9, molecular formula C6H10O4, molecular weight 146.14 g/mol, and listed purity 99.82%.
  • Listed purity of 99.82%.
  • Crystalline solid form for stable storage and handling.
  • Synthetic intermediate for organic and polymer chemistry.
  • Suitable as an analytical standard and reagent in research.
  • Available in multiple pack sizes, including 500 g.
Catalog No. 50-003-01783 Supplier Medchemexpress LLC Supplier No. HYW017522500G
Adipic acid (hexanedioic acid) is a straight-chain dicarboxylic acid supplied as a crystalline solid for laboratory use. It is commonly used as a reagent, chemical intermediate, and analytical standard; manufacturer listings report CAS 124-04-9, molecular formula C6H10O4, molecular weight 146.14 g/mol, and listed purity 99.82%.
  • Listed purity of 99.82%.
  • Crystalline solid form for stable storage and handling.
  • Synthetic intermediate for organic and polymer chemistry.
  • Suitable as an analytical standard and reagent in research.
  • Available in multiple pack sizes, including 500 g.