Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adipic Acid, Solstice

Catalog No. 6002251 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-022-51 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-022-51 Supplier Solstice Supplier No. 2763525KG

missing translation for 'validMainframeMsg'

Meets analytical specification of Ph.Eur., BP, E 355, 99.6-101.0% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-04-9
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00004420
InChI Key WNLRTRBMVRJNCN-UHFFFAOYSA-N
Synonym adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german
PubChem CID 196
ChEBI CHEBI:30832
IUPAC Name hexanedioic acid
SMILES C(CCC(=O)O)CC(=O)O

Specifications

Quantity 25 kg
Flash Point 196°C (384.8°F)
Ignition Point 788°F
Residue after Ignition ≤0.002% (as SO4)
Linear Formula HOOC(CH2)4COOH
Vapor Density 5 (vs air)
Vapor Pressure 1mmHg (159.5°C)
Beilstein 1209788
Solubility Information Freely soluble in methanol, boiling water, cyclohexane, acetone, water, ethanol. Insoluble in benzene,petroleum ether.
Formula Weight 146.14g/mol
Percent Purity 99.6 to 101% (calc. to the dried substance)
Chemical Name or Material Adipic Acid, Meets analytical specification of Ph.Eur.BP
Show More Show Less
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 204-673-3
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.