Learn More
Adipic acid monoethyl ester, 95%
Description
Adipic acid monoethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adipic acid monoethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 28°C to 29°C |
| Density | 0.98 |
| Boiling Point | 179°C to 181°C (decomposition) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.439 |
| Quantity | 5 g |
| Solubility Information | Soluble in water. |
| Formula Weight | 174.2 |
| Percent Purity | 95% |
| Chemical Name or Material | Adipic acid monoethyl ester |
Safety and Handling
| EINECSNumber | 210-966-7 |
| RTECSNumber | AV2030000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.