Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adipic acid monoethyl ester, 95%

Catalog No. p-7027333
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2400606 5 g
AAB2400614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2400606 Supplier Thermo Scientific Chemicals Supplier No. B2400606
Only null left
Add to Cart

CAS: 626-86-8 | C8H14O4 | 174.196 g/mol

Adipic acid monoethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adipic acid monoethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-86-8
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00004419
InChI Key UZNLHJCCGYKCIL-UHFFFAOYSA-N
Synonym adipic acid monoethyl ester, monoethyl adipate, ethyl hydrogen adipate, hexanedioic acid, monoethyl ester, monoethyl hexanedioate, monoethyladipic acid ester, adipic acid, monoethyl ester, mono-ethyl adipate, hexanedioic acid, 1-ethyl ester, hexanoic acid, monoethyl ester
PubChem CID 12295
IUPAC Name 6-ethoxy-6-oxohexanoic acid
SMILES CCOC(=O)CCCCC(=O)O

Specifications

Melting Point 28°C to 29°C
Density 0.98
Boiling Point 179°C to 181°C (decomposition)
Flash Point >110°C (230°F)
Refractive Index 1.439
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 174.2
Percent Purity 95%
Chemical Name or Material Adipic acid monoethyl ester
EINECSNumber 210-966-7
RTECSNumber AV2030000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.